Learn More
Sartorius Cubis II Pipette Calibration System
Product Code. 31662882 Shop All Sartorius Products
Change view
Click to view available options
Product Type:
Pipette Calibration System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31662882 Pipette Calibration System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 31662882 Supplier Sartorius Supplier No. MPS116S
Expédition estimée: 26-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
  • MPS116S is a Cubis™ II-based Single Channel Pipette Calibration System that combines high resolution and large weighing capacity.
  • Together with the option to reduce readability to 10 or 100 g, the instrument is capable of covering up to 95% of the volume ranges for pipettes in the market without sacrificing measurement speed.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipette Calibration System