Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Primer Pairs
Product Code. 15419192 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10,000 pmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419192 10,000 pmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15419192 Supplier Applied Biosystems™ Supplier No. 450056
 Restricted Item
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Custom 5' Fluorescent Labeled/Unlabeled Pairs
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
TRUSTED_SUSTAINABILITY
Specifications
Description Custom 5' Fluorescent Labeled/Unlabeled Pairs, 10000pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, 12 Months Shelf Life, –20°C Storage Temperature, Desalted Purification, Solution or Dried Down Format, For Use in Fragment Analysis, Gene Expression Studies, Genotyping and PCR
Product Type Custom 5' Fluorescent Labeled or Unlabeled Pairs
Storage Buffer TE or Water
Content And Storage -20°C
Primer 5' Labeled
Quantity 10,000 pmol
For Use With (Application) Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Form Solution or Dried down
For Research Use Only. Not for use in diagnostic procedures.