Learn More
Ambion™ Custom mirVana™ miRNA Inhibitor
Product Code. 15581124
Change view
Click to view available options
Quantity:
250 nmol
5 nmol
Unit Size:
250nmol
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15581124 5 nmol Each
15551511 5 nmol Each
15501134 250 nmol 250nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15581124 Supplier Ambion™ Supplier No. 4464086
 Restricted Item
Expédition estimée: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Custom mirVana™ miRNA Inhibitor
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation. Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description mirVana Custom miRNA Inhibitor, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type mirVana Custom miRNA Inhibitor
Quantity 5 nmol
For Research Use Only. Not for use in diagnostic procedures.