Learn More
Cyclohexylamine Hydrochloride, 98%, BLD Pharm™
Product Code. 31004704
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31004704 500 g 500g
30999432 25 g 25g
30995595 1 kg 1kg
30996354 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31004704 Supplier BLD Pharm Supplier No. BD122790500g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 4998-76-9
Molecular Formula C6H14ClN
MDL Number MFCD00050540
Quantity 500 g
SMILES NC1CCCCC1.[H]Cl
Molecular Weight (g/mol) 135.64
Percent Purity 98%
Chemical Name or Material Cyclohexylamine Hydrochloride