Learn More
Hellma™ Cylindrical cell far UV quartz 5mm light path 1400μL Hellma
Product Code. 12701705
Change view
Click to view available options
Lightpath:
5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lightpath Unit Size
12701705 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12701705 Supplier Hellma™ Supplier No. 120540
  • ONLINE EXCLUSIVE PRICE
    284.03€
    253.00€ / 1 pièces
    Économisez 31.03€
    10% de réduction
Expédition estimée: 02-12-2026