Learn More
Chem-Tainer Low-Profile Cylindrical Tank, PE
Product Code. 11786138 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
45 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11786138 45 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11786138 Supplier Chem-Tainer Supplier No. 0692560
  • ONLINE EXCLUSIVE PRICE
    260.00€
    245.23€ / 1 pièces
    Économisez 14.77€
    5% de réduction
Expédition estimée: 01-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Low-profile design of the PE Cylindrical Tank allows for convenient mixing and filling.
  • Resins are FDA compliant.
  • The self supporting, translucent PE tanks have excellent chemical and impact resistance.
  • Polyethylene covers with a flexible PP hinge and stainless steel (SS) rivets are sold separately.
Specifications
Wall Thickness 47.6 cm
Diameter (Metric) Outer 381 mm
Type Tank
Material Polyethylene
Capacity (English) 12 gal.
Capacity (Metric) 45 L
Height (English) 16 in.
Height (Metric) 40.6 cm
No. per Pack 1 Ea.