Learn More
D-α-Hydroxyglutaric acid disodium, MedChemExpress
Product Code. 16472606
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472606 25 mg 25mg
16459636 50 mg 50mg
16433078 10 mg 10mg
16414376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472606 Supplier Medchem Express Supplier No. HY100542
 Restricted Item
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress D-α-Hydroxyglutaric acid disodium (Disodium (R)-2-hydroxyglutarate) is the principal metabolite accumulating in neurometabolic disease D-2-hydroxyglutaric aciduria.
D-α-Hydroxyglutaric acid disodium is a weak competitive antagonist of α-ketoglutarate (α-KG) and inhibits multiple α-KG-dependent dioxygenases with a Ki of 10.87 mM. D-α-Hydroxyglutaric acid disodium increases reactive oxygen species (ROS) production. D-α-Hydroxyglutaric acid disodium binds and inhibits ATP synthase and inhibits mTOR signaling.
Specifications
Chemical Name or Material D-α-Hydroxyglutaric acid disodium
Physical Form Powder
CAS 103404-90-6
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C5H6Na2O5
For Use With (Application) Cancer-Kinase/protease
Synonym Disodium (R)-2-hydroxyglutarate
Solubility Information H2O : 75 mg/mL (390.46 mM; Need ultrasonic)
SMILES O=C(O[Na])[C@H](O)CCC(O[Na])=O
Molecular Weight (g/mol) 192.08
Formula Weight 192.08
Percent Purity 95.0%
Grade Research