missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol, 91.0 to 100.5% (Dry Basis), Ultrapure

Product Code. 15835338 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15835338

Brand: Thermo Scientific J21710.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products

This item is not returnable. View return policy

CAS: 50-70-4 | C6H14O6 | 182.172 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2 R,3 R,4 R,5 S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 98°C to 100°C
Packaging Plastic bottle
Quantity 1 kg
Beilstein 1721899
Formula Weight 182.17
Percent Purity 91.0 to 100.5%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material D-Sorbitol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.