Learn More
Decitabine, MedChemExpress
Product Code. 16487976
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487976 100 mg 100mg
16428258 10 mg 10mg
16404147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487976 Supplier Medchem Express Supplier No. HYA0004
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
Specifications
Chemical Name or Material Decitabine
CAS 2353-33-5
Color White
Purity Grade Notes Research
Molecular Formula C8H12N4O4
Quantity 100 mg
Synonym 5-Aza-2'-deoxycytidine 5-AZA-CdR NSC 127716
Solubility Information DMSO : ≥ 50 mg/mL (219.10 mM) ∣H2O : 20 mg/mL (87.64 mM; Need ultrasonic)
SMILES O=C1N=C(N)N=CN1[C@H](O[C@@H]2CO)C[C@@H]2O
Molecular Weight (g/mol) 228.21
Formula Weight 228.21
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death