Learn More
Delta2-Ceftazidime (>80%), TRC
Product Code. 30009310
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009310 1 mg 1mg
30003862 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009310 Supplier Toronto Research Chemicals Supplier No. TRCC2441101MG
 Restricted Item
Expédition estimée: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1000980-60-8
Synonym Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-en-3-yl]methyl]-, inner salt∣Ceftazidime Delta-3-Isomer
Specifications
Format Neat
Molecular Weight (g/mol) 546.58
Product Type Impurity
Quantity 1 mg
Shipping Condition Room Temperature