Learn More
Deoxy Donepezil Hydrochloride, TRC
Product Code. 30364601
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364601 50 mg 50mg
30360425 25 mg 25mg
30352848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364601 Supplier Toronto Research Chemicals Supplier No. TRCD23262550MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1034439-57-0
Molecular Formula C24 H31 N O2 . Cl H
Molecular Weight (g/mol) 401.97
IUPAC Name 1-benzyl-4-[(5,6-dimethoxy-2,3-dihydro-1H-inden-2-yl)methyl]piperidine;hydrochloride
SMILES Cl.COc1cc2CC(CC3CCN(Cc4ccccc4)CC3)Cc2cc1OC
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H31NO2.ClH/c1-26-23-15-21-13-20(14-22(21)16-24(23)27-2)12-18-8-10-25(11-9-18)17-19-6-4-3-5-7-19;/h3-7,15-16,18,20H,8-14,17H2,1-2H3;1H
Formula Weight 401.2122
Percent Purity >95%
Chemical Name or Material Deoxy Donepezil Hydrochloride
Grade HPLC