Learn More
Honeywell KCL™ Dermatril™ 740 Gloves
Click to view available options
Size:
6
8
9
10
missing translation for 'unitSize'
Pack of 100
Descrizione
- Disposable
- Chemically resistant in accordance with EN374
- High protection against chemical and bacteriological contamination
- Very good mechanical durability in comparison to conventional disposable gloves
- Approved for food handling and processing according to EU 1935/2004
- Free from natural latex
- Resistant against a multitude of cytostatic drugs virus-resistant according to ASTM 1671:2007
- Glove surface is silicone free
- Meets requirements of EN 455
- Medical device according to 93/42/EWG
- Excellent sense of touch
- Extremely comfortable
- Increased tear resistance
- Roughened fingertips for improved grip
- Snug fit
Specifica
Specifica
| Certifications/Compliance | ASTM 1671 2007 |
| Certifications/Compliance (Europe) | EC Category PPE 3, EN374, EU 1935/2004, EN 455, 93/42/EWG |
| Color | Blue |
| Material | Nitrile |
| Powdered | Powder Free |
| Size | 9 |
| Thickness (Metric) | 0.11 mm |
| Length (Metric) | 240 to 260 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction