Learn More
Desoxycarbadox, TRC
Product Code. 30010474
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010474 10 mg 10mg
30132531 25 mg 25mg
30003245 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010474 Supplier Toronto Research Chemicals Supplier No. TRCD29699510MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55456-55-8
Molecular Formula C11 H10 N4 O2
Molecular Weight (g/mol) 230.2227
Synonym Hydrazinecarboxylic acid, (2-quinoxalinylmethylene)-, methyl ester (9CI), Desoxycarbadox, Methyl 3-(2-quinoxalinylmethylene)carbazate
IUPAC Name methyl N-[(E)-quinoxalin-2-ylmethylideneamino]carbamate
SMILES COC(=O)N\N=C\c1cnc2ccccc2n1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C11H10N4O2/c1-17-11(16)15-13-7-8-6-12-9-4-2-3-5-10(9)14-8/h2-7H,1H3,(H,15,16)/b13-7+
Formula Weight 230.0804
Percent Purity >95%
Chemical Name or Material Desoxycarbadox
Grade HPLC