Learn More
Cole-Parmer™ Detachable Electrode Cable
Product Code. 15301122 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
3 m
Unit Size:
10ft
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15301122 3 m 10ft
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15301122 Supplier Cole-Parmer™ Supplier No. 0566256
  • ONLINE EXCLUSIVE PRICE
    95.00€
    88.04€ / 10ft
    Économisez 6.96€
    7% de réduction
Expédition estimée: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Features BNC connection for compatibility with most pH meters and controllers
  • Electrode cable, detachable, 10-ft, BNC
  • Compatible with most pH meters and controllers
Specifications
Type Cole-Parmer Electrode Cable, Detachable, 10-ft, BNC
Includes BNC Connection
Length (English) 10 ft.
Length (Metric) 3 m