missing translation for 'onlineSavingsMsg'
Learn More

Deuterium Oxide 99.90% D, Eurisotop

Product Code. 15287843
Encompass
Change view
Click to view available options
Deuterium Oxide; CAS: 7789-20-0; Deuterium (D): 99.90% D; Grade: Research Grade; Color: Colorless; Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15287843 Deuterium Oxide; CAS: 7789-20-0; Deuterium (D): 99.90% D; Grade: Research Grade; Color: Colorless; Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15287843 Supplier Eurisotop Supplier No. D214K
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Synonym: Heavy Water

Deuterium Oxide

  • Isotopic Enrichment: 99.90% D
  • Quantity: 500 mL

CoA and MSDS available at Eurisotop.com

Specifications

Color Colorless
Deuterium (D) 99.90% D
Quantity 500 mL
CAS 7789-20-0
Melting Point 3.81°C
Boiling Point 101.42°C
Molecular Formula H2O
MDL Number MFCD00044636
Synonym Heavy Water
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES [2H]O[2H]
IUPAC Name [(²H)oxy](²H)
Molecular Weight (g/mol) 20.03
Grade Research Grade
Chemical Name or Material Deuterium Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.