Learn More
Scotch-Brite™ Diamond Floor Pads, Purple
Product Code. 17108800
Change view
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
685 mm
Unit Size:
Pack of 5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
17108800 530 mm Pack of 5
17158800 254 mm Pack of 5
17128800 280 mm Pack of 5
17118800 685 mm Pack of 5
17148800 305 mm Pack of 5
17138800 355 mm Pack of 5
17158790 380 mm Pack of 5
17148790 406 mm Pack of 5
17168790 432 mm Pack of 5
17178790 460 mm Pack of 5
17188790 480 mm Pack of 5
17198790 505 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 17108800 Supplier Scotch-Brite™ Supplier No. 7100159503
  • ONLINE EXCLUSIVE PRICE
    607.00€
    362.09€ / 5 pièces
    Économisez 244.91€
    40% de réduction
Expédition estimée: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Diamond-coated floor pads for cleaning and polishing marble, terrazzo, slate, and polished concrete flooring. Helps eliminate floor finish and reduce labor costs.
  • Simultaneously cleans and polishes flooring to achieve an outstanding shine
  • Ideal for marble, terrazzo, vinyl, slate, and polished concrete floors
  • Easy to use: no expensive or harmful chemicals needed, only water
  • Recommended for use with low and standard speed machines
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Pads
Color Purple
Diameter (Metric) 530 mm
For Use With (Application) Burnishing, Cleaning
For Use With (Equipment) Marble, Terrazzo, Slate, and Polished Concrete Flooring
Material Diamond-coated
Style Round