Learn More
Bochem™ Diamond Pencil with Octahedron Point
Product Code. 15610249
Change view
Click to view available options
Length (Metric):
135 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15610249 135 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15610249 Supplier Bochem™ Supplier No. 12621
  • ONLINE EXCLUSIVE PRICE
    80.50€
    62.22€ / 1 pièces
    Économisez 18.28€
    22% de réduction
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Bochem™ diamond pencil is used for permanent labelling of microscope slides and other glass products in the lab. Etched glass is resistant to humidity and washing.
  • Permanent labelling of glassware and microscope slides
  • Octahedron point
  • Steel handle
Specifications
Handle Material Steel
Length (Metric) 135 mm
Description Diamond Pencil
For Use With (Application) Permanent Marking Glass Microscope Slides and Other Lab Glassware
Tip Style Octahedron Point