Learn More
Dibutyl adipate, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 105-99-7 |
| Molecular Formula | C14H26O4 |
| MDL Number | MFCD00009451 |
| Quantity | 25 g |
| SMILES | O=C(OCCCC)CCCCC(OCCCC)=O |
| Molecular Weight (g/mol) | 258.35 |
| Percent Purity | 99% |
| Chemical Name or Material | Dibutyl adipate |