Learn More
Dichlorobenzene-1.2 D4 99.00% D, Eurisotop
Product Code. 17237456
Encompass_Preferred
Change view
Click to view available options
Dichlorobenzene-1; CAS: 2199-69-1; Deuterium (D): 99.00% D; Grade: Research Grade; Color: Colorless; Quantity: 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17237456 Dichlorobenzene-1; CAS: 2199-69-1; Deuterium (D): 99.00% D; Grade: Research Grade; Color: Colorless; Quantity: 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17237456 Supplier Eurisotop Supplier No. D220FD
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Synonym: 1,2-Dichlorobenzene-d4, Tetradeutero-1,2-dichlorobenzene
1,2-Dichlorobenzene D4
  • Isotopic Enrichment: 98.00% D
  • Quantity: 5 mL
  • Water Content: water <0.01%
CoA and MSDS available at Eurisotop.com
Specifications
Color Colorless
Deuterium (D) 99.00% D
Quantity 5 mL
CAS 2199-69-1
Boiling Point 178 to 180°C
Molecular Formula C6H4Cl2
Synonym 1,2-Dichlorobenzene-d4, Tetradeutero-1,2-dichlorobenzene
InChI Key RFFLAFLAYFXFSW-RHQRLBAQSA-N
SMILES [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
IUPAC Name dichloro(²H₄)benzene
Molecular Weight (g/mol) 151.02
Grade Research Grade
Chemical Name or Material 1.2-Dichlorobenzene D4