missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-aminoethyl)phosphonate oxalate, 97%

Product Code. 10135683 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10135683

Brand: Thermo Scientific Acros 332220050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 203192-98-7 | C6H17NO3P | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203192-98-7
Molecular Formula C6H17NO3P
Molecular Weight (g/mol) 182.18
MDL Number MFCD00800256
InChI Key JUAGHBASZWRMQH-UHFFFAOYSA-O
Synonym diethyl 2-aminoethyl phosphonate oxalate, diethyl aminoethyl phosphonate oxalate, 2-diethoxyphosphoryl ethylamine oxalate, 2-diethoxyphosphorylethanamine; oxalic acid, oxalic acid diethyl 2-aminoethylphosphonate, diethyl 2-aminoethyl phosphonate oxalate salt, diethyl 2-aminoethyl phosphonate ethanedioate, aminoethylphosphonic acid diethylester oxalate salt
PubChem CID 2733595
IUPAC Name 2-diethoxyphosphorylethanamine;oxalic acid
SMILES CCOP(=O)(CC[NH3+])OCC

Specifications

Melting Point 135.0°C to 141.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 271.21
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material Diethyl (2-aminoethyl)phosphonate oxalate
compliance-icons
Product Identifier
  • Diethyl (2-aminoethyl)phosphonate oxalate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.