Learn More
Diethyl Chlorothiophosphate-d10, TRC
Product Code. 30378518
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378518 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30378518 Supplier Toronto Research Chemicals Supplier No. TRCD4441621MG
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diethyl-d10 Chlorothiophosphate
CAS 287397-89-1
Molecular Formula C4 D10 Cl O2 P S
Quantity 1 mg
Synonym Phosphorochloridothioic Acid O,O-(Diethyl-d10) Ester, O,O-(Diethyl-d10) Thiophosphoryl Chloride, NSC 43776-d10
InChI Formula InChI=1S/C4H10ClO2PS/c1-3-6-8(5,9)7-4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])OP(=S)(Cl)OC([2H])([2H])C([2H])([2H])[2H]
IUPAC Name chloro-bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 198.67
Formula Weight 198.0455