Learn More
Diethyl Methylmalonate, TRC
Product Code. 30379932
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379932 50 g -
30378067 5 g 5g
30388131 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379932 Supplier Toronto Research Chemicals Supplier No. TRCD44443050G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 609-08-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
Synonym 2-Methylpropanedioic Acid 1,3-Diethyl Ester, Methylmalonic Acid Diethyl Ester, Diethyl 2-Methylmalonate, Diethyl Methylpropanedioate, Diethyl alpha-Methylmalonate, NSC 8700
SMILES CCOC(=O)C(C)C(=O)OCC
Specifications
Quantity 50 g
InChI Formula InChI=1S/C8H14O4/c1-4-11-7(9)6(3)8(10)12-5-2/h6H,4-5H2,1-3H3
Formula Weight 174.0892
Chemical Name or Material Diethyl Methylmalonate