Learn More
Digi-Sense™ Precalibrated CFM/CMM Vane Thermoanemometer
Product Code. 15860026
Change view
Click to view available options
Airflow Rate:
80 to 5900 fpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
15860026 80 to 5900 fpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15860026 Supplier Digi-Sense™ Supplier No. 2025014
Expédition estimée: 15-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Includes
vane probe sensor with 3.9-ft (1.2-m) cable, three AAA batteries, and NIST-traceable calibration report with data supplied by InnoCal.
High precision with extra large backlit LCD and flow area data storage
  • Reliable measurements guaranteed
  • Precalibrated to NIST-traceable standards
  • User-selectable units of measure eliminate the need for conversion
  • Measure air velocities up to 5900 ft/min (30 m/s) to cover a broad range of applications
  • Measure air flow up to 999,900 CFM, 999,900 CMM
  • Two memory locations, one CFM and one CMM, store commonly used vent area sizes to save you time
  • Large dual-function 4-digit LCD with backlight for easy reading
  • Auto power-off extends battery life
  • Fits comfortably in your hand
Warranty
2 Years
Specifications
Type Vane Thermoanemometer
Accuracy ±3% +40 ft./min. or 0.2 m/s
Airflow Rate 80 to 5900 fpm
Case Material Plastic
Gas Type Air
Color Blue, Gray
Description CFM/CMM Vane Thermoanemometer with NIST Traceable Calibration
For Use With (Application) HVAC or outdoor environments
Height (English) 1 in.
Height (Metric) 2.54 cm
Length (English) 6.75 in.
Length (Metric) 17.1 cm
Power Supply Details Three AAA batteries
Width (Metric) 6.4 cm
Temperature (English) 32°F to 122°F
Temperature (Metric) 0°C to 50°C
Width (English) 2.5 in.