Description
Applied Biosystems Diomni Software is an advanced, browser-based application designed to digitally integrate real-time PCR (qPCR) systems. It supports the planning of an extraction run, running of a qPCR test, reviewing of interpretive results and QC checks, and releasing of reports. Diomni Software can be deployed on a local network to enable centralized data management and role-based access.
Purchase includes an Enterprise license that unlocks additional features. This license must be renewed at the end of the licensing period to maintain access to the Enterprise features.
The software is US-IVD and IVDR compliant.
Features of Diomni Software include:
- Advanced results interpretation capabilities
- Comprehensive automated quality control (QC) checks
- Integrated sample and reagent tracking library
- Connectivity to laboratory information systems (LIS) and laboratory information management systems (LIMS) for efficient test order reception and result transmission
- Connect and start runs on the QuantStudio 7 Pro Dx and QuantStudio 5 Dx real-time PCR systems and receive results automatically after run completion
- Interface with KingFisher Apex Dx Sample Purification System to send extraction runs and receive status updates in real time
- Scalable user management supporting an unlimited number of users
- Enhanced data management and workflow optimization tools, including automated QC checks, sample tracking, and LIS/LIMS integration
Advanced run templates for standardized testing
Diomni Software offers advanced run templates (ADF) that significantly enhance standardization and reduce user errors across multiple sites and laboratories. These templates allow for the locking of test parameters, ensuring consistency among various users.
The software supports a wide range of applications, including presence/absence, standard curve, relative quantification, melt, and genotyping. Furthermore, the system allows for the configuration of results interpretation rules, enabling precise sample result determination. Advanced options such as delta Cq for grouped targets in presence/absence tests, definition of Cq ranges for grey zones, and quality control settings based on curve quality provide users with comprehensive analytical capabilities.
Centralized installation for efficient fleet management
The Diomni Software installation process is designed for optimal efficiency and accessibility. A one-time on-site setup in a central location on a local area network (LAN) enables seamless fleet management. This configuration allows users to initiate qPCR runs on multiple networked instruments simultaneously. Notably, there is no limitation on the number of networked users who can access the shared database across the lab network, facilitating collaborative research and streamlined operations.
Comprehensive workflow support
Diomni Software provides extensive workflow support, guiding users through every stage of the experimental process. The software assists in planning extraction runs, executing qPCR tests, reviewing interpretive results and quality control checks, and generating and releasing reports. This end-to-end support ensures a smooth and efficient experimental workflow, reducing the potential for errors and increasing overall productivity.
Enhanced functionality with internet connectivity
While Diomni Software operates effectively without an internet connection, additional features become available when connected. These include access to a reagent database for document downloads, automated license renewal, and the ability to contact remote technical support through Smart Help. For a comprehensive overview of configuration options, users are encouraged to refer to the connectivity brochure. This flexibility allows laboratories to tailor their setup according to their specific needs and security requirements.
Specifications
Specifications
| Software Compatibility | QuantStudio 7 Pro Dx and QuantStudio 5 Dx, Windows 11 or Windows 10, SAE Administrator Console v3 or SAE Administrator Console Dx v1.2 |
| Product Type | Software |
| License | 3 Years |
| Quantity | 1 Ea. |
For In Vitro Diagnostic Use