Learn More
Direct Blue 218 (Technical Grade), TRC
Product Code. 18050320
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18050320 1 g 1g
18011820 5 g 5g
18010820 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18050320 Supplier Toronto Research Chemicals Supplier No. TRCD4929701G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28407-37-6
Molecular Formula C32 H18 Cu2 N6 O16 S4 . 4 Na
Molecular Weight (g/mol) 1087.81
Synonym 1-Naphthol-3,6-disulfonic acid, 2,2'-(3,3'-dihydroxy-4,4'-biphenylylenebisazo)bis[8-amino-, dicopper deriv., tetrasodium salt (6CI), C.I. Direct Blue 218 (8CI), Copper, [tetrahydrogen-3,3'-[(3,3'-dihydroxy-4,4'-biphenylylene)bis(azo)]bis[5-amino-4-hydroxy-2,7-naphthalenedisulfonato](4-)]di-, tetrasodium salt (7CI), Cuprate(4-), [μ-[[3,3'-[(3,3'-dihydroxy[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[5-amino-4-hydroxy-2,7-naphthalenedisulfonato]](8-)]]di-, tetrasodium, Cuprate(4-), [μ-[[3,3'-[[3,3'-di(hydroxy-κO)[1,1'-biphenyl]-4,4'-diyl]bis(azo-κN1)]bis[5-amino-4-(hydroxy-κO)-2,7-naphthalenedisulfonato]](8-)]]di-, tetrasodium (9CI), 2,7-Naphthalenedisulfonic acid, 3,3'-[(3,3'-dihydroxy[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[5-amino-4-hydroxy-, copper complex, Amanil Supra Blue 9GL, C.I. 24401, Carta Blue VP, Direct Blue 218, Fastusol Blue 9GLP, Intralite Blue 8GLL, Pontamine Bond Blue B, Pontamine Fast Blue 7GLN
SMILES [Na+].[Na+].[Na+].[Na+].Nc1cc(cc2cc(c3c(c12)[O-][Cu+2]4[N]3=Nc5ccc(cc5[O-]4)c6ccc7N=[N]8[Cu+2]([O-]c7c6)[O-]c9c8c(cc%10cc(cc(N)c9%10)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C32H24N6O16S4.2Cu.4Na/c33-19-11-17(55(43,44)45)5-15-9-25(57(49,50)51)29(31(41)27(15)19)37-35-21-3-1-13(7-23(21)39)14-2-4-22(24(40)8-14)36-38-30-26(58(52,53)54)10-16-6-18(56(46,47)48)12-20(34)28(16)32(30)42;;;;;;/h1-12,39-42H,33-34H2,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;;;/q;2*+2;4*+1/p-8
Formula Weight 1085.7688
Chemical Name or Material Direct Blue 218