Learn More
BD Discardit™ Non-Sterile Two-Part Syringe
Product Code. 10286173
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
Graduations:
0.2 mL
0.5 mL
Unit Size:
Pack of 1700
Pack of 3000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Unit Size
10286173 10 mL 0.5 mL Pack of 1700
10012022 5 mL 0.2 mL Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For pressure applications, a three-part syringe is recommended.
Product Code. 10286173 Supplier BD Supplier No. 301360
  • ONLINE EXCLUSIVE PRICE
    496.00€
    322.34€ / 1700 pièces
    Économisez 173.66€
    35% de réduction
En stock
Add to basket
Edge
Add to basket
For pressure applications, a three-part syringe is recommended.
Used for injection or aspiration of medical fluids, including corporal fluids (blood, etc) and drugs. BD™ Discardit™ Non-Sterile Two-Part Syringe is a medical single-use syringe with lubricant in the barrel material for smooth advancement of the plunger without the need of a rubber ring.
  • Two-piece syringes with Luer pip and leak-tight design
  • Clear barrel with accurate graduation scale markings for perfect visualization of contents
  • Ergonomically designed plunger for single-handed operation with reduced sliding force
  • Silicone oil-free lubricant enables smooth, gradual plunger movement
  • Retaining ring prevents accidental plunger rod withdrawal
  • Design of finger grip flanges ensure stability and comfort during injection
  • Single use, individually wrapped syringes are latex- and PVC-free
TRUSTED_SUSTAINABILITY
Specifications
Material Oleamide (Lubricant), Polyethylene (Plunger), Polypropylene (Barrel)
Color Clear
For Use With (Application) Injection and/or aspiration of medical fluids
Product Type 2-part Syringe
Disposable Single-use
Graduations 0.5 mL
Quantity 100/Pk.
Volume (Metric) 10 mL
Connection Type Luer Tip
Sterility Non-sterile