Learn More
Bochem™ Disposable Aluminum Dish
Product Code. 10479962
Change view
Click to view available options
Capacity (Metric):
110 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10479962 110 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10479962 Supplier Bochem™ Supplier No. 11611
  • ONLINE EXCLUSIVE PRICE
    47.03€
    35.58€ / 100 pièces
    Économisez 11.45€
    24% de réduction
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Manufactured from aluminum. BOCHEM™ Disposable Aluminum Dish are disposable.
  • Disposable
  • Manufactured from aluminum
Specifications
Bottom Shape Flat
Capacity (Metric) 110 mL
Disposable Single-use
Material Aluminum
Shape Round
Type Dish
Height (Metric) 25 mm
Diameter (Metric) 99 mm