Learn More
Velp Scientifica™ DK 6 Digester
Product Code. 17134185 Shop All Velp Products
Change view
Click to view available options
Capacity (Metric):
6 x 250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17134185 6 x 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17134185 Supplier Velp Scientifica™ Supplier No. F30100400
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Kjeldahl Digestion Units with a 6-position aluminum heating block that offers an excellent thermal homogeneity.
  • The DK 6 Series digestion unit with aluminum heating block ensures high thermal homogeneity heating up to 450°C. The temperature of the block is constantly controlled by a microprocessor and is extremely stable, homogeneous and precise: ±0.5°C at all selectable temperatures.
  • The DK 6 Series guarantees accuracy and repeatability. The DK 6 series perform batch digestion of up to 6 samples with 250 mL test tubes - 42 mm dia., reducing the energy consumption (-13% compared to the previous series). Low energy consumption provides lower costs of analysis and limits CO2 release.
  • The design of the DK series has been carefully engineered to allow laboratories to save valuable space under the hood.
  • DK Digesters have very compact dimensions: the new aluminum heating block has been lowered and the vertical proportions allow to free precious space in the laboratory.
  • The structure is made of stainless steel that has been specifically designed and tested to ensure strong resistance to chemicals and mechanical stress.
  • The aluminum block protection grid provides enhanced heating performance and protects the heating block from splashes and chemicals.
  • The bright digital display provides maximum visibility and easy-to-read information.
  • The instrument is operated with 4 keys to navigate the menu options and program the digestions in a simple and fast way.
  • Through the intuitive graphic display it is possible to set up to 20 methods with 4 temperature ramps.
  • Acoustic signals inform the operator of the completion of the analysis facilitating the daily use of the digester.
  • The inclination of the front panel protects it from chemical spillage at high temperatures.
  • DK 6 is a versatile digester suitable for Kjeldahl N/Protein applications and Chemical Oxygen Demand in the environmental industry.
    • In order to use the DK 6 for COD (Chemical Oxygen Demand) is necessary to configure it by adding a different support system able to manage all the glass required for the COD analysis for processing 6 samples simultaneously.
  • The DK 6 and TMD 6 are the reliable solution for digestion with aqua regia (nitrohydrochloric acid) in order to determine the acid-soluble proportion of metals in sewage sludges, sediments and soils In accordance with official methods (DIN 38414-S7 standard).
  • For application where the digestion process produces fumes, acid gases or corrosive substances, the below specific accessories are strongly recommended:
    • JP Pump aspirates noxious fumes
    • SMS Scrubber neutralizes all the fumes
Specifications
Display Digital
Voltage 230 V
Capacity (English) 6 x 8.5 oz.
Capacity (Metric) 6 x 250 mL
Dimensions (L x W x H) 130 x 350 x 220 mm
Includes Instrument Only
Material Stainless Steel, Epoxy Painted with Chemical Resistant Coating
No. of Places 6
Temperature (English) Maximum 842°F
Temperature (Metric) Maximum 450°C
Frequency 50/60 Hz