Learn More
asecos™ Document Holder
Product Code. 11988837 Shop All asecos Products
Change view
Click to view available options
For Use With (Equipment):
All Cabinets 590 to 1555 mm Wide
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11988837 All Cabinets 590 to 1555 mm Wide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11988837 Supplier asecos™ Supplier No. 30793
  • ONLINE EXCLUSIVE PRICE
    50.96€
    37.00€ / 1 pièces
    Économisez 13.96€
    27% de réduction
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Polystyrene document holder, DIN A5
Specifications
Description Document holder DIN A5
For Use With (Equipment) All Cabinets 590 to 1555 mm Wide
Weight (Metric) 0.30 kg
Material Plastic
Product Type Document holder