Learn More
Hanna Instruments™ Dosing Pump
Product Code. 11850164
Change view
Click to view available options
Flow Rate:
1.5 L/hr.
10 L/hr.
20 L/hr.
5 L/hr.
7 L/hr.
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
11850164 1.5 L/hr. Each
10601742 7 L/hr. Each
10345721 5 L/hr. Each
10274131 10 L/hr. Each
10458822 20 L/hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11850164 Supplier Hanna Instruments™ Supplier No. BL1.52
  • ONLINE EXCLUSIVE PRICE
    593.55€
    344.65€ / 1 pièces
    Économisez 248.90€
    41% de réduction
View all promotions
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Overall Height (Metric) 150 mm
Overall Length (Metric) 180 mm
Temperature (Metric) Max 45°C
Flow Rate 1.5 L/hr.
Maximum Pressure 12 bar
Weight (Metric) 4 kg