Learn More
Dräger™ CMS Chips: Chlorine
Product Code. 10076001
Change view
Click to view available options
Measurement Range:
0.2 to 10
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10076001 0.2 to 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10076001 Supplier Dräger™ Supplier No. 6406010
  • ONLINE EXCLUSIVE PRICE
    230.36€
    191.70€ / 1 pièces
    Économisez 38.66€
    16% de réduction
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Chlorine CMS Chip
Humidity Range 5 to 12mg/L
Measurement Range 0.2 to 10
Response Time 30 to 400 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C