Learn More
Dräger™ CMS Chips: Benzene
Product Code. 12344609
Change view
Click to view available options
Measurement Range:
0.2 to 10
0.5 to 10
10 to 250
Response Time:
35 to 225 sec.
35 to 300 sec.
40 to 275 sec.
Humidity Range:
1 to 30mg/L
1 to 40mg/L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time Humidity Range Unit Size
12344609 10 to 250 40 to 275 sec. 1 to 30mg/L Each
10233972 0.5 to 10 35 to 225 sec. 1 to 40mg/L Each
10709772 0.2 to 10 35 to 300 sec. 1 to 30mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12344609 Supplier Dräger™ Supplier No. 6406280
  • ONLINE EXCLUSIVE PRICE
    260.55€
    203.28€ / 1 pièces
    Économisez 57.27€
    21% de réduction
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Benzene CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 250
Response Time 40 to 275 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C