Learn More
Dräger™ CMS Chips: Oxygen
Product Code. 10786695
Change view
Click to view available options
Measurement Range:
1 to 30%
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10786695 1 to 30% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10786695 Supplier Dräger™ Supplier No. 6406490
  • ONLINE EXCLUSIVE PRICE
    411.52€
    321.80€ / 1 pièces
    Économisez 89.72€
    21% de réduction
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Air Pressure 700 to 1100 hpa
Type Oxygen CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 1 to 30%
Response Time 100 to 600 sec.
Temperature Range (English) 33.8°F to 104°F
Temperature Range (Metric) 1°C to 40°C