Learn More
Dräger™ X-act™ 5000 Automatic Tube Pump: Chargers
Product Code. 13537270
Change view
Click to view available options
Type:
Wall Charger
Dimensions (L x W x H):
6 x 6 x 6 in. (15.2 x 15.2 x 15.2cm)
Voltage:
110/230VAC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Dimensions (L x W x H) Voltage Unit Size
13537270 Wall Charger 6 x 6 x 6 in. (15.2 x 15.2 x 15.2cm) 110/230VAC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13537270 Supplier Dräger™ Supplier No. 4523545
  • ONLINE EXCLUSIVE PRICE
    149.85€
    117.30€ / 1 pièces
    Économisez 32.55€
    21% de réduction
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Dimensions (L x W x H) 6 x 6 x 6 in. (15.2 x 15.2 x 15.2cm)
Type Wall Charger
Height (English) 6 in.
Height (Metric) 15.2 cm
Length (English) 6 in.
Length (Metric) 15.2 cm
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C
Voltage 110/230VAC
Width (English) 6 in.
Width (Metric) 15.2 cm