Learn More
Dräger™ XS-2 Sensors for MiniWarn™ Multi-Gas Monitors
Product Code. 11322080
Change view
Click to view available options
Measurement Range:
0 to 25%
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
11322080 0 to 25% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11322080 Supplier Dräger™ Supplier No. 6810375
  • ONLINE EXCLUSIVE PRICE
    782.78€
    591.32€ / 1 pièces
    Économisez 191.46€
    24% de réduction
 Restricted Item
Expédition estimée: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Detecting or monitoring oxygen gas
  • Measuring range of 0-25% volume
  • 0.1% volume resolution
  • Compatible with Pac III, MiniWarn and Multiwarn II monitors
Warranty
2 years
Specifications
Components Measured Oxygen
Humidity Range 10 to 95%
Measurement Range 0 to 25%
Pressure Range 700 to 1300mbar
Resolution 0.1
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 2 Years