Learn More
Neogen Dry-Sponge with Glove BP237SPG
Product Code. 17248530
Encompass
Change view
Click to view available options
Dimensions (L x W):
1.5 x 3 in.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17248530 1.5 x 3 in. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17248530 Supplier Neogen Supplier No. 7000042939
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Biocide-free cellulose sponge that includes two polyethylene gloves and a sample bag.
  • 1.5 x 3 in. dry sponge can be used with a wide range buffers and enrichment media and in dry-only environments.
  • Simplified packaging convenience, including sponge, sample bag and gloves, minimizes handling and preparation and reduces waste.
  • 5 x 9 in. sample bag with 18 oz./0.53 L capacity and included for safe handling and convenient transportation to lab.
  • Biocide-free sponge does not affect organism viability.
  • Decontaminated through gamma irradiation to maintain aseptic conditions.
Specifications
Product Type Dry Sponge
Dimensions (L x W) 1.5 x 3 in.
Packaging Quantity 100/Cs.