Learn More
Lenz Laborglasintrumente Drying Tubes, Bent, (Calcium Chloride Tubes)
Product Code. 10356261
Change view
Click to view available options
Length (Metric):
100 mm
150 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10356261 150 mm Each
10010221 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356261 Supplier Lenz Laborglasintrumente Supplier No. 05333013
  • ONLINE EXCLUSIVE PRICE
    12.25€
    10.07€ / 1 pièces
    Économisez 2.18€
    17% de réduction
Expédition estimée: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Drying tubes acc. to DIN 12610, bent
  • With ball and hose connection
  • Made of DURAN™ tubing
Specifications
Length (Metric) 150 mm
Material Borosilicate Glass (3.3)