Learn More
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground
Product Code. 15884518 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
14.5 NS
18.8 NS
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
15884518 18.8 NS Pack of 50
15864518 14.5 NS Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884518 Supplier DWK Life Sciences Supplier No. 286037308
  • ONLINE EXCLUSIVE PRICE
    1350.00€
    1210.50€ / 50 pièces
    Économisez 139.50€
    10% de réduction
Expédition estimée: 29-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
2-way tap plug with thread.
Ground DURAN™ patent tap plugs have 2 almost parallel inclined bores.
  • With thread, ground
  • 2-way tap plug with 2 almost parallel inclined holes
  • massive tap plug
Specifications
Plug Bore 4 mm
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Closure Size 18.8 NS
No. per Pack 25/Pk.
For Use With (Application) Installation in glass apparatus and devices