Learn More
(E)-4-((4-(4-(1H-1,2,3-Triazol-1-yl)butyl)phenoxy)methyl)-2-(4-(trifluoromethyl)styryl)oxazole, 98%, BLD Pharm™
Product Code. 30988627
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30988627 25 mg -
30987430 5 mg -
30984493 50 mg -
30987246 100 mg 100mg
30971084 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30988627 Supplier BLD Pharm Supplier No. BD14052925mg
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 366017-09-6
Molecular Formula C25H23F3N4O2
MDL Number MFCD09954135
Quantity 25 mg
SMILES FC(F)(C1=CC=C(/C=C/C2=NC(COC3=CC=C(C=C3)CCCCN4C=CN=N4)=CO2)C=C1)F
Molecular Weight (g/mol) 468.47
Percent Purity 98%
Chemical Name or Material (E)-4-((4-(4-(1H-1,2,3-Triazol-1-yl)butyl)phenoxy)methyl)-2-(4-(trifluoromethyl)styryl)oxazole