Learn More
Econazole Nitrate, LoGiCal
Product Code. 30458814
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458814 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458814 Supplier LoGiCal Supplier No. LGCFOR0524.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Econazole Nitrate
CAS 24169-02-6
Analyte or Component Names Econazole Nitrate
Shipping Condition Room Temperature
Molecular Formula C18 H15 Cl3 N2 O . H N O3
Certifications/Compliance ISO 9001
Synonym Econazole Nitrate, 1-[(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. B (EP) as Nitrate, 1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole nitrate, Econazole nitrate, Ecostatin, Epi-Pevaryl, Gyno-Pevaril, Gynoryl, Ifenec, Micofugal, Micogin, NSC 243115, Palavale, Pargin, Pevaryl, R 14827, SQ 13050, Spectazole, (+/-)-1-[2-[(4-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole mononitrate, 1-[2,4-Dichloro-beta-[(p-chlorobenzyl)oxy]phenethyl]imidazole mononitrate, (+/-)-Econazole nitrate, Miconazole Impurity B as Nitrate, Miconazole Nitrate Impurity B as Nitrate, Miconazole Impurity B as Nitrate
InChI Formula InChI=1S/C18H15Cl3N2O.HNO3/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21;2-1(3)4/h1-9,12,18H,10-11H2;(H,2,3,4)
Solution Type Neat
SMILES O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 444.7
Quantity 10 mg
Formula Weight 443.0206 g/mol