missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-216 Thermal Work Gloves

Product Code. 18916920 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
Unit Size:
6 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18916920 Universal 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18916920 Supplier Ansell Supplier No. 48216
  • ONLINE EXCLUSIVE PRICE
    80.40€
    61.54€ / 6 paires
    Économisez 18.86€
    23% de réduction
Expédition estimée: 28-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Robust and dexterous thermal work gloves provide heat, flame, spark, and puncture protection.

  • Heightened defenses: REACH compliant and CE-certified. Gloves offer EN 12477 Type A protection for demanding welding and thermal job tasks, ranging from stick and MIG welding to torch cutting and grinding.
  • Enhanced features: Featuring an improved three-dimensional thumb design, high-dexterity gloves offer tactile sensitivity for precise welding control.
  • Increased protection and durability: Heat-resistant gloves boast higher durability, made of premium split cowhide leather and have welted seams sewn with aramid.
  • Added comfort: Breathable work gloves possess a cotton lining that grants thermal insulation and extended comfort.
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Grade Ungraded
Hand Specific
Material Leather
Product Type Welding Gloves
Size Universal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.