Learn More
PL BioScience ELAREM™ Perform, Human Platelet Lysate, Heparin Required
Product Code. 17977731
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Purity or Quality Grade:
GMP Grade
Research Grade
Unit Size:
100mL
500mL
50mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Purity or Quality Grade Unit Size
17977731 50 mL Research Grade 50mL
17987731 50 mL GMP Grade 50mL
17967731 100 mL Research Grade 100mL
17957741 100 mL GMP Grade 100mL
17987721 500 mL Research Grade 500mL
17977721 500 mL GMP Grade 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17977731 Supplier PL BioScience Supplier No. PE10611
  • ONLINE EXCLUSIVE PRICE
    245.74€
    200.00€ / 50mL
    Économisez 45.74€
    18% de réduction
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
ELAREM™ Perform is a human Platelet Lysate of EU origin supporting in vitro expansion of various primary cells and cell lines. The safe and performance-increasing cell growth promoter ensures efficient lab processes with batch-to-batch consistency and a xeno-free cell environment.
  • Xeno-Free: The xeno-free cell culture supplement is obtained from transfusion-approved platelets obtained at licensed blood centres.
  • EU Origin: ELAREM™ Perform Research Grade is manufactured from platelet units obtained from healthy blood donors at licensed blood centres in the EU.
  • Improved Cell Performance: Human Platelet Lysate contains abundant growth factors and cytokines derived from human platelets. Those nutrients have been shown to stimulate cellular proliferation and maintain phenotype and differentiation potential of various cells.
  • Reproducible Results: Each batch of ELAREM™ Perform is produced from large pools of transfusion-approved platelet units to ensure batch-to-batch consistency and enable reproducible conditions. Grade & GMP GradeB
  • From Research to Clinical Development: ELAREM™ Perform meets the requirements of academic and industrial cell expansion and assures an easy transition of research results into clinical applications.
  • The product contains coagulation factors. To inhibit coagulation of the complete cell culture medium, it is recommended to add an anticoagulant, e.g. heparin, to the cell culture medium.
  • Intended Use: ELAREM™ Perform is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).
Specifications
Color Clear, Yellow
Culture Type Adherent Cell Culture, Suspension Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile
Sterilization Method Sterile-filtered
Formulation Liquid, Human Platelet Lysate
Purity or Quality Grade Research Grade
Quantity 50 mL
Shipping Condition Dry Ice