Learn More
IKA™ ElectraSyn Complete Electrode and Vial Starter Kit
Product Code. 15877922 Shop All IKA Products
Change view
Click to view available options
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15877922 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877922 Supplier IKA™ Supplier No. 0040004013
Expédition estimée: 26-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Accessories kit that includes cap assemblies, all vial sizes, and all electrode types for the ElectraSyn 2.0 multi tool.
  • Includes sets of vials in all sizes:
    • Vial 5 mL with complete cap assembly
    • Vial 10 mL with complete cap assembly
    • Vial 20 mL with complete cap assembly
    • Vial 5 mL set of 10
    • Vial 10 mL set of 10
    • Vial 20 mL set of 20
  • Includes sets of all electrode types
    • Graphite SK-50, Set of 12
    • Glassy carbon, Set of 2
    • Platinum plated, Set of 2
    • Magnesium, Set of 12
    • Zinc, Set of 12
    • Stainless steel, Set of 12
    • Nickel foam, Set of 12
    • Nickel, Set of 12
    • RVC, Set of 12
TRUSTED_SUSTAINABILITY
Specifications
Description Complete vial, cap, and electrode set.