Learn More
Corning™ Elplasia™ 48K Vessel
Product Code. 31932523 Shop All Corning Products
Change view
Click to view available options
Shape:
Rectangular
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Shape Unit Size
31932523 Rectangular Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31932523 Supplier Corning™ Supplier No. 9920
Expédition estimée: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Enables researchers to generate a high density of spheroids in a scaffold-free model.
  • The Corning™ Elplasia™ 48K Vessel enables researchers to generate a high density of spheroids in a scaffold free model
  • The Corning Elplasia 48K vessel has a 320 cm2 surface area with 152 microcavities per cm2 and generates approximately 48000 spheroids of uniform size and shape per vessel 4X the output of the Elplasia 12K Flask and Open Well Plate
  • Gravity in conjunction with the Corning Ultra Low Attachment ULA surface and a rounded microcavity geometry enable formation of approximately 48000 spheroids of similar shape and size
  • This is a custom made to order product
  • Please contact us for additional information and details on ordering
TRUSTED_SUSTAINABILITY
Specifications
Shape Rectangular
Surface Coating Ultra-Low Attachment
Capacity (Metric) 48,000 spheroids of similar shape and size
Diameter (Metric) Outer Microcavity: 850 μm; Maximum Spheroid: 400 μm
Material Polystyrene