Learn More
Merck Flow-limiting Orifices
Product Code. 10182111
Change view
Click to view available options
Airflow Rate:
1 L/min.
2 L/min.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
10182111 2 L/min. Each
10072423 1 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182111 Supplier Merck Supplier No. XX5000020
  • ONLINE EXCLUSIVE PRICE
    149.00€
    143.00€ / 1 pièces
    Économisez 6.00€
    4% de réduction
Expédition estimée: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Creates quantitative airflow through a filter for airborne contamination analysis
Flow Limiting Orifices create quantitative airflow through a MF-Millipore filter for airborne contamination analysis. The orifices are threaded into the outlet of a Millipore Aerosol Analysis Holder or Aerosol Adapter to enable a constant flow rate when required vacuum level is applied.Ratings are accurate to within approximately 15%. For greater precision, calibrate orifice with a flowmeter at actual working vacuum level.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Environmental Test Kits or Supplies
Material Stainless Steel
For Use With (Equipment) Millipore Aerosol Analysis Holder or Aerosol Adapter
For Use With (Application) To create quantitative airflow through a Millipore filter for airborne contamination analysis
Airflow Rate 2 L/min.
Includes Flow-limiting Orifice, 2 L/min