Learn More
Merck Histosec™ Pastilles Embedding Media
Product Code. 17888321
Change view
Click to view available options
Quantity:
4 x 2.5kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17888321 4 x 2.5kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17888321 Supplier Merck Supplier No. 1116092504
 Restricted Item
Expédition estimée: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
solidification point 56-58°C embedding agent for histology
Histosec™ pastilles is an embedding medium for histology of samples of human origin. It is a paraffin enriched with polymers, also available without the addition of DMSO (dimethyl sulfoxide). DMSO increases the rate of penetration of paraffin and provides additional preservation of the tissue structures. Its melting range is between 56 - 58 °C and, within this range any specimen can be processed without fear of damage irrespective of the method used.
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 54° to 57°C
Product Type Pastilles
Quantity 4 x 2.5kg
For Use With (Application) Histology