missing translation for 'onlineSavingsMsg'
Learn More

ENDOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22148-1-AP-150UL

Nacres classification code: NA.46

 View more versions of this product

Product Code. 16851685

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
Specifications

Specifications

ENDOG
Polyclonal
Unconjugated
ENDOG
Endo G, ENDOG, endonuclease G, Endonuclease G, mitochondrial
Rabbit
Antigen Affinity Chromatography
RUO
13804, 2021, 362100
-20°C
Liquid
Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08600, Q14249
Endog
ENDOG Fusion Protein Ag17739
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.