Learn More
Corning™ Endothelial Cell Growth Supplement (ECGS)
Product Code. 11553520 Shop All Corning Products
Change view
Click to view available options
Quantity:
15 mg
100 mg
Unit Size:
100mg
15mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11553520 15 mg 15mg
11523610 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11553520 Supplier Corning™ Supplier No. 354006
  • ONLINE EXCLUSIVE PRICE
    154.84€
    138.35€ / 15mg
    Économisez 16.49€
    10% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Ideal for endothelial cell culture
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Product Type Growth Supplement
Quantity 15 mg