Learn More
Ertugliflozin, MedChemExpress
Product Code. 16496538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496538 10 mg 10mg
16429127 5 mg 5mg
16422637 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496538 Supplier Medchem Express Supplier No. HY15461
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Specifications
Chemical Name or Material Ertugliflozin
CAS 1210344-57-2
Color White
Purity Grade Notes Research
Molecular Formula C22H25ClO7
Quantity 10 mg
Synonym PF-04971729
Solubility Information DMSO : 250 mg/mL (572.24 mM; Need ultrasonic)
SMILES ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4
Molecular Weight (g/mol) 436.88
Formula Weight 436.88
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism