Learn More
Estradiol, MedChemExpress
Product Code. 16462059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462059 Supplier Medchem Express Supplier No. HYB0141
 Restricted Item
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Estradiol (β-Estradiol) is a steroid hormone and the major female sex hormone. Estradiol can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. Estradiol can be used for the research of cancers, neurodegenerative diseases and neural tissue engineering.
Estradiol (β-Estradiol) is a steroid hormone and the major female sex hormone. Estradiol can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. Estradiol can be used for the research of cancers, neurodegenerative diseases and neural tissue engineering.
Specifications
Chemical Name or Material Estradiol
CAS 50-28-2
Color White
Purity Grade Notes Research
Molecular Formula C18H24O2
Quantity 1 g
Synonym β-Estradiol E2 17β-Estradiol 17β-Oestradiol
Solubility Information DMSO : ≥ 50 mg/mL (183.57 mM) ∣Ethanol : 20 mg/mL (73.43 mM; Need ultrasonic)
SMILES C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 272.38
Formula Weight 272.38
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death